Potassium trichloroammineplatinate(II)
Potassium trichloroammineplatinate(II)
Potassium trichloroammineplatinate(II)
Potassium trichloroammineplatinate(II)
Thermo Scientific Chemicals

Potassium trichloroammineplatinate(II)

CAS: 13820-91-2 | Cl3H3KNPt | 357.563 g/mol
Have Questions?
Catalog NumberQuantity
039433.ME
also known as 039433-ME
500 mg
Catalog number 039433.ME
also known as 039433-ME
Price (EUR)
464,95
Online Exclusive
561,00
Save 96,05 (17%)
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
464,95
Online Exclusive
561,00
Save 96,05 (17%)
Each
Add to cart
Chemical Identifiers
CAS13820-91-2
IUPAC Namepotassium trichloroplatinumuide amine
Molecular FormulaCl3H3KNPt
InChI KeyPIFYEDQIQOTXLO-UHFFFAOYSA-K
SMILESN.[K+].Cl[Pt-](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormCrystals or crystalline powder
Assay from Suppliers CofAChloride: ≥28.2%
Assay from Suppliers CofAPlatinum: ≥51.8%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only