Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis)
Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis)
Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis)

CAS: 126956-48-7 | C2NiO4 | 146.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 040757.14
also known as 040757-14
Price (EUR)
196,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
196,00
Each
Chemical Identifiers
CAS126956-48-7
IUPAC Namenickel(2+) oxalate
Molecular FormulaC2NiO4
InChI KeyDOLZKNFSRCEOFV-UHFFFAOYSA-L
SMILES[Ni++].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0015% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only