Cobalt(II) phthalocyanine
Cobalt(II) phthalocyanine
Cobalt(II) phthalocyanine
Cobalt(II) phthalocyanine
Thermo Scientific Chemicals

Cobalt(II) phthalocyanine

CAS: 3317-67-7 | C32H16CoN8 | 571.469 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 041496.06
also known as 041496-06
Price (EUR)
107,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
107,00
Each
Add to cart
Chemical Identifiers
CAS3317-67-7
IUPAC Nameλ²-cobalt(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Molecular FormulaC32H16CoN8
InChI KeyMPMSMUBQXQALQI-UHFFFAOYSA-N
SMILES[Co++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple
Formcrystalline powder
Assay (unspecified)> 95.0%
Cobalt(II) phthalocyanine may be used to develop carbon black based electrocatalysts. CoPC may be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalyzed aerobic regenerations of aldehydes and ketones have been investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only