Di-n-octyl phthalate, 98%
Di-n-octyl phthalate, 98%
Di-n-octyl phthalate, 98%
Thermo Scientific Chemicals

Di-n-octyl phthalate, 98%

Plasticizer | CAS: 117-84-0 | C24H38O4 | 390.564 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Catalog number 041522.AC
also known as 041522-AC
Price (EUR)
63,10
Each
Quantity:
25 mL
Request bulk or custom format
Price (EUR)
63,10
Each
Specifications
Assay Percent Range98%
CAS117-84-0
ChEBICHEBI:34679
Chemical Name or MaterialDi-n-octyl phthalate
Density0.98g/mL
View more
Di-n-octyl phthalate is used as a plasticizer for many resins and elastomers. It acts as an additive. It is also used for medical tubing and blood storage bags, wire and cables, carpet back coating, floor tile and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only