Methanol-d{4}, 99.8% (Isotopic)
Methanol-d{4}, 99.8% (Isotopic)
Methanol-d{4}, 99.8% (Isotopic)
Methanol-d{4}, 99.8% (Isotopic)
Thermo Scientific Chemicals

Methanol-d{4}, 99.8% (Isotopic)

CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 Ea.
2 Ea.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042318.KV
also known as 042318-KV
Price (EUR)
-
Quantity:
10 Ea.
Request bulk or custom format
Chemical Identifiers
CAS811-98-3
IUPAC Name(²H₃)methan(²H)ol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES[2H]OC([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay by NMR>/= 99.0%
Formliquid
Appearance (Color)Colorless
Methyl alcohol-d4 is the labeled analog of methyl alcohol, a common laboratory solvent. It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only