n-Hexane, anhydrous
n-Hexane, anhydrous
n-Hexane, anhydrous
Thermo Scientific Chemicals

n-Hexane, anhydrous

Solvent in Sol-Gel Precursor Applications | CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 043263.AK
also known as 043263-AK
Price (EUR)
59,90
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
59,90
Each
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Residue on Evaporation≤5ppm
Impurity contentSulfur compounds (as Sulfur) ≤0.005%
Water Content (Karl Fischer Titration)≤0.02%
Assay (GC)≥99.5% (as C6 Hydrocarbons)
View more
n-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only