Fullerene, nanotube, multi-walled, 20 nm OD, 5-20 micron long
Thermo Scientific Chemicals

Fullerene, nanotube, multi-walled, 20 nm OD, 5-20 micron long

CAS: 308068-56-6 | CH4 | 16.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Catalog number 043839.MF
also known as 043839-MF
Price (EUR)
43,50
Each
Quantity:
50 mg
Request bulk or custom format
Price (EUR)
43,50
Each
Specifications
CAS308068-56-6
ChEBICHEBI:16183
Chemical Name or MaterialFullerene
ColorBlack
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only