Potassium hexabromopalladate(IV), Premion™, 99.999% (metals basis), Pd 15.5% min
Potassium hexabromopalladate(IV), Premion™, 99.999% (metals basis), Pd 15.5% min
Potassium hexabromopalladate(IV), Premion™, 99.999% (metals basis), Pd 15.5% min
Thermo Scientific Chemicals

Potassium hexabromopalladate(IV), Premion™, 99.999% (metals basis), Pd 15.5% min

CAS: 16919-74-7 | Br6K2Pd | 664.041 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043947.06
also known as 043947-06
Price (EUR)
440,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
440,00
Each
Chemical Identifiers
CAS16919-74-7
IUPAC Namedipotassium tetrabromopalladium dibromide
Molecular FormulaBr6K2Pd
InChI KeySKCNWISEAGYZFC-UHFFFAOYSA-H
SMILES[K+].[K+].[Br-].[Br-].Br[Pd](Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only