4-Methyl-3-nitrophenol, 98%
4-Methyl-3-nitrophenol, 98%
4-Methyl-3-nitrophenol, 98%
4-Methyl-3-nitrophenol, 98%
Thermo Scientific Chemicals

4-Methyl-3-nitrophenol, 98%

CAS: 2042-14-0 | C7H7NO3 | 153.137 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044285.18
also known as 044285-18
Price (EUR)
207,00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
207,00
Each
Add to cart
Chemical Identifiers
CAS2042-14-0
IUPAC Name4-methyl-3-nitrophenol
Molecular FormulaC7H7NO3
InChI KeyBQEXDUKMTVYBRK-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)Yellow
Assay (GC)> 97.0%
It is used as a chemical, pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only