Aluminum chloride, anhydrous, 99.999% (metals basis)
Aluminum chloride, anhydrous, 99.999% (metals basis)
Aluminum chloride, anhydrous, 99.999% (metals basis)
Thermo Scientific Chemicals

Aluminum chloride, anhydrous, 99.999% (metals basis)

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 044470.06
also known as 044470-06
Price (EUR)
84,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
84,50
Each
Chemical Identifiers
CAS7446-70-0
IUPAC Namealuminium(3+) trichloride
Molecular FormulaAlCl3
InChI KeyVSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Residual water< 2000 ppm
Total Metal Impurities≤0.001% (10 ppm)
Particle Size-10 Mesh
FormCrystalline
It is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only