Cadmium sulfide, 99.999% (metals basis)
Cadmium sulfide, 99.999% (metals basis)
Cadmium sulfide, 99.999% (metals basis)
Cadmium sulfide, 99.999% (metals basis)
Thermo Scientific Chemicals

Cadmium sulfide, 99.999% (metals basis)

Used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. | CAS: 1306-23-6 | CdS | 144.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045455.18
also known as 045455-18
Price (EUR)
177,00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
177,00
Each
Add to cart
Chemical Identifiers
CAS1306-23-6
IUPAC Namecadmium(2+) sulfanediide
Molecular FormulaCdS
InChI KeyFRLJSGOEGLARCA-UHFFFAOYSA-N
SMILES[S--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Cadmium sulfide is used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. It is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only