Thermo Scientific Chemicals

Amberlite™ IRN-78, ion exchange resin, nuclear grade, Thermo Scientific Chemicals

Catalog number: 045591.36
500 g, Each
Thermo Scientific Chemicals

Amberlite™ IRN-78, ion exchange resin, nuclear grade, Thermo Scientific Chemicals

Catalog number: 045591.36
500 g, Each
Quantity
Catalog number: 045591.36
also known as 045591-36
Price (EUR)
Price: 49,20
Online price: 44,28
Your price:
Quantity
-

Chemical Identifiers

CAS
11128-95-3
MDL Number
MFCD00145822
Form
Spherical beads
Moisture holding capacity
≥54.0 to ≤60.0% (as Hydroxide form)
Appearance (Color)
White to yellow to amber
Comment
Coefficient of Uniformity: ≤1.20
Harmonic mean size
≥0.580 to ≤0.680mm

Description

Amberlite™ IRN-78 is widely employed in removing radioactive anions for example 131 Iodine and 133 Iodine from waste systems. It also removes the traces of chloride contamination from reactor coolant systems and controls the boron level in the primary system. At higher pH conditions, it is used to remove anionic impurities from secondary streams in the presence of ammonia, morpholine or other amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H318 - Causes serious eye damage

    Precautionary Statements

    P280 - Wear eye protection/ face protection

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician