3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392
3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392
3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392
Thermo Scientific Chemicals

3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392

CAS: 1071466-61-9 | C25H28O4 | 392.495 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 046403.14
also known as 046403-14
Price (EUR)
60,90
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
60,90
Each
Specifications
CAS1071466-61-9
Chemical Name or Material3,3'-Diallylbisphenol A diacetate
ColorAmber
Formula Weight392.5
Health Hazard 1H315-H319-H335
View more
It is used in epoxy systems or in combination with acrylates, methacrylates, and maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Recommended for use in epoxy systems or in combination with acrylates, methacrylates, and maleimides
RUO – Research Use Only