Cobalt phosphide, 99.9%
Cobalt phosphide, 99.9%
Thermo Scientific Chemicals

Cobalt phosphide, 99.9%

CAS: 12134-02-0 | Co{2}P
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 047333.06
also known as 047333-06
Price (EUR)
101,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
101,00
Each
Add to cart
Chemical Identifiers
CAS12134-02-0
IUPAC Nametris(λ²-cobalt(2+)) diphosphanetriide
Molecular FormulaCo3P2
InChI KeyYYHKCTWKZVGLSL-UHFFFAOYSA-N
SMILES[P-3].[P-3].[Co++].[Co++].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
FormPowder
Assay from Supplier's CofA≥99.9%
Particle Sizeca. 100 Mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.