Strontium iodide hexahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium iodide hexahydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number 047367.22
also known as 047367-22
Price (EUR)
84,70
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
84,70
Each
Specifications
CAS7790-40-1
Melting Point90°C (decomposition)
Molecular FormulaSrI{2}^0.6 H{2}O
Percent Purity99%
Physical FormSolid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.