Sodium hydroxide, 30% w/w aq. soln.
Sodium hydroxide, 30% w/w aq. soln.
Sodium hydroxide, 30% w/w aq. soln.
Thermo Scientific Chemicals

Sodium hydroxide, 30% w/w aq. soln.

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 087864.AP
also known as 087864-AP
Price (EUR)
67,70
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
67,70
Each
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
ConcentrationSodium hydroxide 30% w/w aq. soln.

Sodium hydroxide, 30% w/v aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing of food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only