Acenaphthenequinone, 95%
Acenaphthenequinone, 95%
Acenaphthenequinone, 95%
Thermo Scientific Chemicals

Acenaphthenequinone, 95%

CAS: 82-86-0 | C12H6O2 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 102191000
Price (EUR)
204,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
204,00
Each
Chemical Identifiers
CAS82-86-0
IUPAC Name1,2-dihydroacenaphthylene-1,2-dione
Molecular FormulaC12H6O2
InChI KeyAFPRJLBZLPBTPZ-UHFFFAOYSA-N
SMILESO=C1C(=O)C2=C3C1=CC=CC3=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to ochre
Infrared spectrumConforms
HPLC>=94.0 %
Appearance (Form)Powder and/or granules
Melting point253°C to 263°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only