2-Acetylbenzoic acid, 99%
2-Acetylbenzoic acid, 99%
2-Acetylbenzoic acid, 99%
Thermo Scientific Chemicals

2-Acetylbenzoic acid, 99%

CAS: 577-56-0 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 102471000
Price (EUR)
275,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
275,00
Each
Chemical Identifiers
CAS577-56-0
IUPAC Name2-acetylbenzoic acid
Molecular FormulaC9H8O3
InChI KeyQDAWXRKTSATEOP-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Appearance (Color)Light yellow to light orange
Melting point115°C to 118°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only