Thermo Scientific Chemicals

4-Biphenylcarboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 106261000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Biphenylcarboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 106261000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 106261000
Price (EUR)
Price: 196,00
Online price: 174,44
Your price:
Quantity
-

Chemical Identifiers

CAS
3218-36-8
Appearance (Form)
Crystalline powder or crystals
Melting point
57°C to 60°C
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell