3-Chlorobenzoyl chloride, 99+%
3-Chlorobenzoyl chloride, 99+%
3-Chlorobenzoyl chloride, 99+%
Thermo Scientific Chemicals

3-Chlorobenzoyl chloride, 99+%

CAS: 618-46-2 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
50 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 108745000
Price (EUR)
497,00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
497,00
Each
Chemical Identifiers
CAS618-46-2
IUPAC Name3-chlorobenzoyl chloride
Molecular FormulaC7H4Cl2O
InChI KeyWHIHIKVIWVIIER-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC=<0.5 % o-Chlorobenzoyl chloride
Appearance (Color)Clear colorless
Infrared spectrumConforms
Titration RCOOH=<1.0 % (as Chlorobenzoic acid)
GC=<0.03 % Total other poly-halogenated acid halides
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only