2-Chlorobenzylamine, 97%
2-Chlorobenzylamine, 97%
2-Chlorobenzylamine, 97%
2-Chlorobenzylamine, 97%
Thermo Scientific Chemicals

2-Chlorobenzylamine, 97%

CAS: 89-97-4 | C7H8ClN | 141.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 108780250
Price (EUR)
42,50
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
42,50
Each
Add to cart
Chemical Identifiers
CAS89-97-4
IUPAC Name1-(2-chlorophenyl)methanamine
Molecular FormulaC7H8ClN
InChI KeyKDDNKZCVYQDGKE-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.5610 to 1.5640 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only