Cytidine, 99+%
Cytidine, 99+%
Cytidine, 99+%
Thermo Scientific Chemicals

Cytidine, 99+%

CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 111810100
Price (EUR)
61,50
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
61,50
Each
Chemical Identifiers
CAS65-46-3
IUPAC Name4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular FormulaC9H13N3O5
InChI KeyUHDGCWIWMRVCDJ-UHFFFAOYNA-N
SMILESNC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrophotometrymolar absorptivity 13000 +/- 2%, on dry basis
Appearance (Form)Crystalline powder
Melting point210°C to 217°C (Dec)
Loss on drying=<0.5 % (105°C, 5 hrs)
UV transmittance>=98 % (c=1, H2O, 430 nm) 1 cm cell
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only