1,3-Indanedione, 97%
1,3-Indanedione, 97%
1,3-Indanedione, 97%
Thermo Scientific Chemicals

1,3-Indanedione, 97%

CAS: 606-23-5 | C9H6O2 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 122090250
Price (EUR)
106,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
106,00
Each
Chemical Identifiers
CAS606-23-5
IUPAC Name2,3-dihydro-1H-indene-1,3-dione
Molecular FormulaC9H6O2
InChI KeyUHKAJLSKXBADFT-UHFFFAOYSA-N
SMILESO=C1CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige to golden
Infrared spectrumConforms
Melting point124°C to 131°C
Titration with NaOH>=96.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only