Thermo Scientific Chemicals

4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: 123450250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: 123450250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 123450250
Price (EUR)
Price: 83,70
Online price: 75,33
Your price:
Quantity
-

Chemical Identifiers

CAS
25026-34-0
Infrared spectrum
Conforms
GC
>=95.0 %
Refractive index
1.5490 to 1.5570 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear yellow to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Skin Sensitization
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H314 - Causes severe skin burns and eye damage

    H317 - May cause an allergic skin reaction

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician