4,4'-Methylenebis(N,N-dimethylaniline), 98%
4,4'-Methylenebis(N,N-dimethylaniline), 98%
4,4'-Methylenebis(N,N-dimethylaniline), 98%
Thermo Scientific Chemicals

4,4'-Methylenebis(N,N-dimethylaniline), 98%

CAS: 101-61-1 | C17H22N2 | 254.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 126745000
Price (EUR)
400,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
400,00
Each
Chemical Identifiers
CAS101-61-1
IUPAC Name4-{[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Molecular FormulaC17H22N2
InChI KeyJNRLEMMIVRBKJE-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(CC2=CC=C(C=C2)N(C)C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey to brown
Infrared spectrumConforms
Melting point86°C to 92°C
Appearance (Form)Powder or crystals and/or chunks
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only