Propionaldehyde, 97%
Propionaldehyde, 97%
Propionaldehyde, 97%
Thermo Scientific Chemicals

Propionaldehyde, 97%

CAS: 123-38-6 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 131510025
Price (EUR)
118,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
118,00
Each
Chemical Identifiers
CAS123-38-6
IUPAC Namepropanal
Molecular FormulaC3H6O
InChI KeyNBBJYMSMWIIQGU-UHFFFAOYSA-N
SMILESCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3610 to 1.3680 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only