Tetraethylthiuram Disulfide, 97%
Tetraethylthiuram Disulfide, 97%
Tetraethylthiuram Disulfide, 97%
Thermo Scientific Chemicals

Tetraethylthiuram Disulfide, 97%

CAS: 97-77-8 | C10H20N2S4 | 296.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Catalog number 138120250
Price (EUR)
20,10
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
20,10
Each
Chemical Identifiers
CAS97-77-8
IUPAC NameN,N-diethyl[(diethylcarbamothioyl)disulfanyl]carbothioamide
Molecular FormulaC10H20N2S4
InChI KeyAUZONCFQVSMFAP-UHFFFAOYSA-N
SMILESCCN(CC)C(=S)SSC(=S)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or granules
Melting point67°C to 74°C
HPLC>=96.0 %
Water=<1.0 % (KF)
Appearance (Color)White to cream to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only