L-Tyrosine, 99+%
L-Tyrosine, 99+%
L-Tyrosine, 99+%
L-Tyrosine, 99+%
Thermo Scientific Chemicals

L-Tyrosine, 99+%

L-Tyrosine, CAS # 60-18-4, is an aromatic non-essential amino acid that is considered a fundamental metabolite, involved in protein synthesis and photosynthesis. | CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 140640025
Price (EUR)
1.007,00
Each
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (EUR)
1.007,00
Each
Chemical Identifiers
CAS60-18-4
IUPAC Name(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO3
InChI KeyOUYCCCASQSFEME-SVGMAFHSNA-N
SMILESN[C@@H](CC1=CC=C(O)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 % (on dried substance)
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Form)Powder
Infrared spectrumConforms
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• L-Tyrosine is the levorotatory isomer of the aromatic amino acid tyrosine, synthesized in vivo from L-phenylalanine
• It is available as a crystalline, off-white powder, stable at room temperature, and soluble in hydrochloric acid

Applications

• In vivo L-tyrosine is involved in protein synthesis and is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catecholamines.
• Used as a supplement, L-tyrosine has been shown to increase levels of the neurotransmitters, dopamine, adrenaline, and norepinephrine in the brain
• It is considered an essential dietary supplement for phenylketonuria patients who lack the enzyme phenylalanine hydroxylase, and hence cannot convert phenylalanine into tyrosine
• L-tyrosine plays a vital role as an electron-carrier in the process of photosynthesis
• Due to its role in the synthesis of catecholamines, L-tyrosine is also used in research to study its impact on stress

RUO – Research Use Only

General References:

  1. Young, S. N. L-Tyrosine to alleviate the effects of stress? J Psychiatry Neurosci. 2007, 32(3), 224.
  2. Hase, A.; Jung, S. E.; Rot, M. Behavioral and cognitive effects of tyrosine intake in healthy human adults. Pharmacol Biochem Behav 2015, 133, 1-6.
  3. Webster, D.; Wildgoose, J. Tyrosine supplementation for phenylketonuria. Cochrane Database Syst Rev.
  4. 2013 Jun 5;2013(6):CD001507.
  5. Barry, B. A.; Babcock, G. T. Tyrosine radicals are involved in the photosynthetic oxygen-evolving system. Proc Natl Acad Sci U S A. 1987, 84(20), 7099-103.