n-Amyl acetate, 99%
n-Amyl acetate, 99%
n-Amyl acetate, 99%
Thermo Scientific Chemicals

n-Amyl acetate, 99%

n-Amyl acetate, 99%, C7H14O2, CAS Number-628-63-7, 1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate, 2.5L, 98.5% min. (GC), 02, 131, 149.0 deg.C, 100.0, CHEBI:87362 | CAS: 628-63-7 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 149180010
Price (EUR)
99,40
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
99,40
Each
Chemical Identifiers
CAS628-63-7
IUPAC Namepentyl acetate
Molecular FormulaC7H14O2
InChI KeyPGMYKACGEOXYJE-UHFFFAOYSA-N
SMILESCCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4010 to 1.4030 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only