2-Methoxyethanol, 99.5+%, for analysis
2-Methoxyethanol, 99.5+%, for analysis
2-Methoxyethanol, 99.5+%, for analysis
2-Methoxyethanol, 99.5+%, for analysis
Thermo Scientific Chemicals

2-Methoxyethanol, 99.5+%, for analysis

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 149360025
Price (EUR)
154,00
Each
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
154,00
Each
Add to cart
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.5 %
Aldehyde (C2H4O)=<0.003 %
Acidity=<0.001 meq/g
Alkalinity=<0.0005 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only