Benzyl chloroformate, 97 wt%, stabilized
Benzyl chloroformate, 97 wt%, stabilized
Benzyl chloroformate, 97 wt%, stabilized
Thermo Scientific Chemicals

Benzyl chloroformate, 97 wt%, stabilized

Benzyl chloroformate, 97 wt%, stabilized, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 178.0 deg.C to 180.0 deg.C, Undesignated, Melting Point: -20.0 deg.C, Molecular Weight: 170.6, Percent Purity: 97 wt%, Assay Percent Range: 96% min. (GC), Beilstein: 06,437 | CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 152940025
Price (EUR)
708,00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
708,00
Each
Chemical Identifiers
CAS501-53-1
IUPAC Namebenzyl carbonochloridate
Molecular FormulaC8H7ClO2
InChI KeyHSDAJNMJOMSNEV-UHFFFAOYSA-N
SMILESClC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoThe material builds progressively benzyl chloride
Additional infoduring storage
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only