Bromoform, 96%, stabilized
Bromoform, 96%, stabilized
Bromoform, 96%, stabilized
Thermo Scientific Chemicals

Bromoform, 96%, stabilized

CAS: 75-25-2 | CHBr3 | 252.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
5 L
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 158200050
Price (EUR)
935,00
Each
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
935,00
Each
Chemical Identifiers
CAS75-25-2
IUPAC Nametribromomethane
Molecular FormulaCHBr3
InChI KeyDIKBFYAXUHHXCS-UHFFFAOYSA-N
SMILESBrC(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=95 %
Infrared spectrumConforms
Stabilizer3 to 5 wt% ethanol
Appearance (Color)Clear colorless to yellow to light orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only