Quinidine, 98%, anhydrous
Quinidine, 98%, anhydrous
Quinidine, 98%, anhydrous
Thermo Scientific Chemicals

Quinidine, 98%, anhydrous

CAS: 56-54-2 | C20H24N2O2 | 324.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 163681000
Price (EUR)
563,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
563,00
Each
Chemical Identifiers
CAS56-54-2
IUPAC Name(S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Molecular FormulaC20H24N2O2
InChI KeyLOUPRKONTZGTKE-LHHVKLHASA-N
SMILESCOC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
RemainderDihydroquinidine
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only