2-Cyclohepten-1-one, 90%
2-Cyclohepten-1-one, 90%
2-Cyclohepten-1-one, 90%
Thermo Scientific Chemicals

2-Cyclohepten-1-one, 90%

CAS: 1121-66-0 | C7H10O | 110.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 171070050
Price (EUR)
256,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
256,00
Each
Chemical Identifiers
CAS1121-66-0
IUPAC Namecyclohept-2-en-1-one
Molecular FormulaC7H10O
InChI KeyWZCRDVTWUYLPTR-UHFFFAOYSA-N
SMILESO=C1CCCCC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=89.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only