Uric acid, +99%, Thermo Scientific Chemicals
Uric acid, +99%, Thermo Scientific Chemicals
Uric acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Uric acid, +99%, Thermo Scientific Chemicals

Uric acid, CAS # 69-93-2, is a common oxopurine metabolite, and a normal component of urine.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 171290250
Price (EUR)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS69-93-2
IUPAC Name2,3,6,7,8,9-hexahydro-1H-purine-2,6,8-trione
Molecular FormulaC5H4N4O3
InChI KeyLEHOTFFKMJEONL-UHFFFAOYSA-N
SMILESO=C1NC2=C(N1)C(=O)NC(=O)N2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Assay>=99.0 % (ex N)
Loss on drying=<0.5 % (105°C, 3 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Uric acid is a purine-based metabolite found in humans, mice and E. coli
• Uric acid can accumulate in excess leading to diseases such as hyperuricemia
• Effects of excess uric acid accumulation include hypertension, arthritis, and renal issues
Applications

• Useful as a biochemical standard
• Suitable for use in the study of hyperuricemia and gout
• Uric acid may also play other roles in cell metabolism and physiology
• Intended for research use only

RUO – Research Use Only

General References:

  1. Chen, M.; Lu, X.; Lu, C.; Shen, N.; Jiang, Y.; Chen, M.; Wu, H. Soluble uric acid increases PDZK1 and ABCG2 expression in human intestinal cell lines via the TLR4-NLRP3 inflammasome and PI3K/Akt signaling pathway. Arthritis Res Ther. 2018 20(1), 20.
  2. Al Abdulmonem, W.; Aljohani, A. S. M.; Alhumaydhi, F. A.; Mousa, A. H. M.; Rasheed, Z. Protective Potential of Uric Acid, Folic Acid, Glutathione and Ascorbic Acid Against the Formation of Toxic Met-Myoglobin. Protein Pept Lett. 2021 28(3), 282-289.