Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D
Thermo Scientific Chemicals

Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D

CAS: 7698-05-7 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
17542050050 gGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 175420500
Price (EUR)
-
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7698-05-7
IUPAC Nameproton chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES[1H+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
D-Enrichment>=99.0 %
Concentration20 to 22 % w/w
Appearance (Form)Solution
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only