4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
Thermo Scientific Chemicals

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 176320250
Price (EUR)
405,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
405,00
Each
Add to cart
Chemical Identifiers
CAS16593-81-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=99 % (on dry substance)
Water=<10 % (K.F.)
Suitability(for Pb determination) Passes test
Solubility(0.1 % in water) Clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only