Ethyl 4-chloroacetoacetate, 98%
Ethyl 4-chloroacetoacetate, 98%
Ethyl 4-chloroacetoacetate, 98%
Ethyl 4-chloroacetoacetate, 98%
Thermo Scientific Chemicals

Ethyl 4-chloroacetoacetate, 98%

CAS: 638-07-3 | C6H9ClO3 | 164.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 177642500
Price (EUR)
92,10
Each
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
92,10
Each
Add to cart
Chemical Identifiers
CAS638-07-3
IUPAC Nameethyl 4-chloro-3-oxobutanoate
Molecular FormulaC6H9ClO3
InChI KeyOHLRLMWUFVDREV-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4515 to 1.4545 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only