Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
Thermo Scientific Chemicals

Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000

CAS: 9003-05-8 | (C3H5NO)n | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Catalog number 178040050
Price (EUR)
34,40
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
34,40
Each
Chemical Identifiers
CAS9003-05-8
Molecular Formula(C3H5NO)n
SMILESNC(=O)C(-*)C-*
MDL NumberMFCD00084392
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<13 %
Appearance (Color)White to light yellow
Appearance (Form)Granules
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only