1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
Thermo Scientific Chemicals

1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene

CAS: 90-13-1 | C10H7Cl | 162.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 180971000
Price (EUR)
40,10
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
40,10
Each
Specifications
Assay Percent Range80% min. (HPLC)
Beilstein05,541
CAS90-13-1
CAS91-58-7
Chemical Name or Material1-Chloronaphthalene
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only