Cobalt(II) chloride hexahydrate, pure
Cobalt(II) chloride hexahydrate, pure
Cobalt(II) chloride hexahydrate, pure
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, pure

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 192860010
Price (EUR)
262,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
262,00
Each
Chemical Identifiers
CAS7791-13-1
IUPAC Nameλ²-cobalt(2+) hexahydrate dichloride
Molecular FormulaCl2CoH12O6
InChI KeyGFHNAMRJFCEERV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to violet
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only