Mercury(II) iodide, ACS reagent, Red
Mercury(II) iodide, ACS reagent, Red
Mercury(II) iodide, ACS reagent, Red
Thermo Scientific Chemicals

Mercury(II) iodide, ACS reagent, Red

CAS: 7774-29-0 | HgI2 | 454.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 193501000
Price (EUR)
94,50
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
94,50
Each
Chemical Identifiers
CAS7774-29-0
IUPAC Namemercury(2+) diiodide
Molecular FormulaHgI2
InChI KeyYFDLHELOZYVNJE-UHFFFAOYSA-L
SMILES[I-].[I-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Titration Iodometric>=99.0 % (Dried basis)
Mercurous Mercury (as Hg)=<0.1 %
Appearance (Form)Crystalline powder
Solubility(In potassium iodide soln.) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only