Betaine, 98%, for analysis, anhydrous
Betaine, 98%, for analysis, anhydrous
Betaine, 98%, for analysis, anhydrous
Thermo Scientific Chemicals

Betaine, 98%, for analysis, anhydrous

CAS: 107-43-7 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 204240050
Price (EUR)
26,65
Online Exclusive
29,20
Save 2,55 (9%)
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
26,65
Online Exclusive
29,20
Save 2,55 (9%)
Each
Chemical Identifiers
CAS107-43-7
IUPAC Name2-(trimethylazaniumyl)acetate
Molecular FormulaC5H11NO2
InChI KeyKWIUHFFTVRNATP-UHFFFAOYSA-N
SMILESC[N+](C)(C)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO497.5 to 102.5 %
Heavy metals (as Pb)=<10 ppm
UV transmittance>=95 % (c=10, H2O, 430 nm) 1 cm cell
Sulfate (SO4)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only