Ammonium hydrogen difluoride, 95%, extra pure, Thermo Scientific Chemicals
Ammonium hydrogen difluoride, 95%, extra pure, Thermo Scientific Chemicals
Ammonium hydrogen difluoride, 95%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hydrogen difluoride, 95%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 208822500
Price (EUR)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1341-49-7
IUPAC Nameamine dihydrofluoride
Molecular FormulaF2H5N
InChI KeyKVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILESN.F.F
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 % (K.F.)
Appearance (Form)Flakes or scales
Iron (Fe)=<500 ppm
Titration with NaOH>=94 %
Impurity=<5 % (Ammonium fluoride (NH4F))
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only