1-Heptadecanol, 97%
1-Heptadecanol, 97%
1-Heptadecanol, 97%
Thermo Scientific Chemicals

1-Heptadecanol, 97%

CAS: 1454-85-9 | C17H36O | 256.474 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 221230050
Price (EUR)
61,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
61,30
Each
Chemical Identifiers
CAS1454-85-9
IUPAC Nameheptadecan-1-ol
Molecular FormulaC17H36O
InChI KeyGOQYKNQRPGWPLP-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point51°C to 56°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only