1-Methyl-2-benzimidazolinone, 98%
1-Methyl-2-benzimidazolinone, 98%
1-Methyl-2-benzimidazolinone, 98%
Thermo Scientific Chemicals

1-Methyl-2-benzimidazolinone, 98%

CAS: 1849-01-0 | C8H8N2O | 148.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2765000505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 276500050
Price (EUR)
152,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
152,00
Each
Chemical Identifiers
CAS1849-01-0
IUPAC Name1-methyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular FormulaC8H8N2O
InChI KeyPYEHNKXDXBNHQQ-UHFFFAOYSA-N
SMILESCN1C(=O)NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to light beige
Melting point192°C to 196°C
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only