Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromosulfuric acid, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 295510025
Price (EUR)
130,00
Each
Request bulk or custom format
Price (EUR)
130,00
Each
Chemical Identifiers
CAS65272-71-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-brown to dark brown
Assay>=1.3 % CrO3
Assay>=92 % H2SO4
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only