Ammonium tetrafluoroborate, 99%, pure
Ammonium tetrafluoroborate, 99%, pure
Ammonium tetrafluoroborate, 99%, pure
Thermo Scientific Chemicals

Ammonium tetrafluoroborate, 99%, pure

CAS: 13826-83-0 | BF4H4N | 104.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 310965000
Price (EUR)
59,00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
59,00
Each
Chemical Identifiers
CAS13826-83-0
IUPAC Nameammonium tetrafluoroboranuide
Molecular FormulaBF4H4N
InChI KeyPDTKOBRZPAIMRD-UHFFFAOYSA-O
SMILES[NH4+].F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only