Sodium peroxide, ACS reagent
Sodium peroxide, ACS reagent
Sodium peroxide, ACS reagent
Thermo Scientific Chemicals

Sodium peroxide, ACS reagent

CAS: 1313-60-6 | Na2O2 | 77.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 315761000
Price (EUR)
99,40
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
99,40
Each
Chemical Identifiers
CAS1313-60-6
IUPAC Namedisodium dioxidanediide
Molecular FormulaNa2O2
InChI KeyPFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Phosphate (PO4)=<5 ppm
AppearanceMay contain a small quantity of discolored
Titration with KMnO4>=93.0 %
Heavy metals (as Pb)=<20 ppm
Sulfate (SO4)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only