Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure
Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure
Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure
Thermo Scientific Chemicals

Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 317230050
Price (EUR)
33,65
線上優惠
36,90
Save 3,25 (9%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
33,65
線上優惠
36,90
Save 3,25 (9%)
Each
Chemical Identifiers
CAS6080-56-4
IUPAC Nameλ²-lead(2+) diacetate trihydrate
Molecular FormulaC4H12O7Pb
InChI KeyMCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White
Total trace metal impurities=<0.001 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only